Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ism2
Entrez Gene ID
100763293
Gene Name
isthmin 2
Gene Aliases
I79_005788, Isthmin-2
Chromosome Location
Chr.Un|NW_003613749: 1764818 - 1781671 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640406.1 XP_007638596.1 2 - 3 600 81

View Full Product Details