Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl22
Entrez Gene ID
100752906
Gene Name
kelch-like family member 22
Gene Aliases
I79_009065
Chromosome Location
Chr.Un|NW_003613898: 1606134 - 1639836 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502462.2 XP_003502510.1 3 - 4 492 67
XM_007643077.1 XP_007641267.1 3 - 4 589 67
GenBank mRNA JI864234.1 - 3 - 4 475 67

View Full Product Details