Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lypd6b
Entrez Gene ID
100768780
Gene Name
LY6/PLAUR domain containing 6B
Gene Aliases
I79_023940
Chromosome Location
Chr.Un|NW_003616833: 76579 - 86030 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514657.1 XP_003514705.1 1 - 2 67 90

View Full Product Details