Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc57
Entrez Gene ID
100771842
Gene Name
coiled-coil domain containing 57
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613637: 3066508 - 3169748 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654255.1 XP_007652445.1 4 - 5 797 79
XM_007654258.1 XP_007652448.1 4 - 5 798 79
XM_007654261.1 XP_007652451.1 4 - 5 798 79
XM_007654267.1 XP_007652457.1 4 - 5 797 79

View Full Product Details