Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shroom1
Entrez Gene ID
103159804
Gene Name
shroom family member 1
Gene Aliases
I79_004326
Chromosome Location
Chr.Un|NW_003613693: 260748 - 279422 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007628930.1 XP_007627120.1 1 - 2 64 63

View Full Product Details