Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prdx6
Entrez Gene ID
100754867
Gene Name
peroxiredoxin 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613658: 1280335 - 1291380 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497679.2 XP_003497727.2 4 - 5 632 96
GenBank mRNA JI875976.1 - 4 - 5 632 96

View Full Product Details