Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnajb1
Entrez Gene ID
100753924
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613865: 1647561 - 1651415 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501888.2 XP_003501936.1 2 - 3 899 102
GenBank mRNA JI872951.1 - 2 - 3 889 102
JP043469.1 - 2 - 3 889 102

View Full Product Details