Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dbx1
Entrez Gene ID
100760100
Gene Name
developing brain homeobox 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613888: 429095 - 433101 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502266.1 XP_003502314.1 3 - 4 667 78

View Full Product Details