Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Abhd16b
Entrez Gene ID
100762917
Gene Name
abhydrolase domain containing 16B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613943: 1461611 - 1463712 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503139.2 XP_003503187.1 1 - 1 1617 71

View Full Product Details