Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cstl1
Entrez Gene ID
100766245
Gene Name
cystatin-like 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614789: 110849 - 115282 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509879.1 XP_003509927.1 1 - 2 226 103

View Full Product Details