Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rai2
Entrez Gene ID
100751572
Gene Name
retinoic acid induced 2
Gene Aliases
I79_011233
Chromosome Location
Chr.Un|NW_003614044: 722215 - 723813 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644740.1 XP_007642930.1 1 - 2 135 106

View Full Product Details