Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Barx2
Entrez Gene ID
100765416
Gene Name
BARX homeobox 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613784: 1787745 - 1852642 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500290.1 XP_003500338.1 2 - 3 489 99

View Full Product Details