Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Grin3b
Entrez Gene ID
100753630
Gene Name
glutamate receptor, ionotropic, N-methyl-D-aspartate 3B
Gene Aliases
I79_009209
Chromosome Location
Chr.Un|NW_003613904: 745297 - 751393 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643247.1 XP_007641437.1 2 - 3 2181 62

View Full Product Details