Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Samd8
Entrez Gene ID
100763843
Gene Name
sterile alpha motif domain containing 8
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613632: 1175578 - 1222397 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652775.1 XP_007650965.1 2 - 3 627 76
GenBank mRNA JI887274.1 - 1 - 2 539 76
JP047645.1 - 1 - 2 539 76

View Full Product Details