Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aqp3
Entrez Gene ID
100758981
Gene Name
aquaporin 3 (Gill blood group)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615295: 212110 - 217751 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511964.2 XP_003512012.1 4 - 5 562 67

View Full Product Details