Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rps19bp1
Entrez Gene ID
100770022
Gene Name
ribosomal protein S19 binding protein 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616334: 68791 - 72039 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514092.2 XP_003514140.1 3 - 4 319 69
GenBank mRNA JI895182.1 - 1 - 2 64 69
JP050784.1 - 1 - 2 64 69

View Full Product Details