Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnajb9
Entrez Gene ID
100763648
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 9
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613674: 2907514 - 2910815 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497996.2 XP_003498044.2 1 - 2 849 94

View Full Product Details