Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tp53tg5
Entrez Gene ID
100764961
Gene Name
TP53 target 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614077: 890354 - 897134 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645115.1 XP_007643305.1 2 - 3 1178 108

View Full Product Details