Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc45a4
Entrez Gene ID
100775003
Gene Name
solute carrier family 45, member 4
Gene Aliases
I79_021681
Chromosome Location
Chr.Un|NW_003615613: 78643 - 108784 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653240.1 XP_007651430.1 4 - 5 1453 63

View Full Product Details