Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gna15
Entrez Gene ID
100761285
Gene Name
guanine nucleotide binding protein (G protein), alpha 15 (Gq class)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614244: 361953 - 384624 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506441.2 XP_003506489.1 2 - 3 333 77
XM_007646956.1 XP_007645146.1 2 - 3 245 77

View Full Product Details