Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tex12
Entrez Gene ID
100769839
Gene Name
testis expressed 12
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613694: 1335704 - 1338763 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007629374.1 XP_007627564.1 3 - 4 572 99

View Full Product Details