Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Msgn1
Entrez Gene ID
100755379
Gene Name
mesogenin 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613936: 739669 - 740235 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503045.1 XP_003503093.1 1 - 1 517 85

View Full Product Details