Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eif3h
Entrez Gene ID
100751998
Gene Name
eukaryotic translation initiation factor 3, subunit H
Gene Aliases
I79_003561
Chromosome Location
Chr.Un|NW_003613668: 840719 - 921518 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007619178.1 XP_007617368.1 6 - 7 685 81
GenBank mRNA JP055479.1 - 4 - 5 693 81

View Full Product Details