Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gtpbp6
Entrez Gene ID
100761234
Gene Name
GTP binding protein 6 (putative)
Gene Aliases
I79_023515
Chromosome Location
Chr.Un|NW_003616509: 65225 - 87991 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654472.1 XP_007652662.1 6 - 7 812 61

View Full Product Details