Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdh18
Entrez Gene ID
100751447
Gene Name
cadherin 18, type 2
Gene Aliases
I79_010174
Chromosome Location
Chr.Un|NW_003613963: 6974 - 272534 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643988.1 XP_007642178.1 4 - 5 808 104

View Full Product Details