Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tslp
Entrez Gene ID
100773425
Gene Name
thymic stromal lymphopoietin
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614043: 815634 - 818193 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644738.1 XP_007642928.1 1 - 2 179 80

View Full Product Details