Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arrdc4
Entrez Gene ID
100770261
Gene Name
arrestin domain containing 4
Gene Aliases
I79_011208
Chromosome Location
Chr.Un|NW_003614042: 155276 - 165444 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504196.1 XP_003504244.1 5 - 6 871 86

View Full Product Details