Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hes2
Entrez Gene ID
100756407
Gene Name
hes family bHLH transcription factor 2
Gene Aliases
I79_024356
Chromosome Location
Chr.Un|NW_003617180: 9406 - 10263 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514972.1 XP_003515020.1 3 - 4 239 90

View Full Product Details