Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cpsf4l
Entrez Gene ID
100762278
Gene Name
cleavage and polyadenylation specific factor 4-like
Gene Aliases
I79_000967
Chromosome Location
Chr.Un|NW_003613598: 2070178 - 2081124 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643271.1 XP_007641461.1 5 - 6 502 87

View Full Product Details