Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcte3
Entrez Gene ID
100774972
Gene Name
t-complex-associated-testis-expressed 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614132: 7555 - 17925 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505267.2 XP_003505315.1 3 - 4 668 76

View Full Product Details