Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kiss1
Entrez Gene ID
100762823
Gene Name
KiSS-1 metastasis-suppressor
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614184: 300391 - 302748 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505744.1 XP_003505792.1 1 - 2 108 92

View Full Product Details