Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptplad1
Entrez Gene ID
100752449
Gene Name
protein tyrosine phosphatase-like A domain containing 1
Gene Aliases
I79_014752
Chromosome Location
Chr.Un|NW_003614337: 194529 - 219839 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647705.1 XP_007645895.1 6 - 7 542 88
GenBank mRNA JI888569.1 - 8 - 9 88

View Full Product Details