Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxl15
Entrez Gene ID
100752453
Gene Name
F-box and leucine-rich repeat protein 15
Gene Aliases
I79_015880
Chromosome Location
Chr.Un|NW_003614474: 575933 - 578037 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648689.1 XP_007646879.1 3 - 4 745 52

View Full Product Details