Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcte1
Entrez Gene ID
100757406
Gene Name
t-complex-associated-testis-expressed 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613855: 1327470 - 1336168 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501676.1 XP_003501724.1 1 - 2 265 90

View Full Product Details