Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nptx2
Entrez Gene ID
100764878
Gene Name
neuronal pentraxin II
Gene Aliases
I79_016015
Chromosome Location
Chr.Un|NW_003614494: 280573 - 296821 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508163.1 XP_003508211.1 1 - 2 79 72

View Full Product Details