Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnk5
Entrez Gene ID
100760223
Gene Name
potassium channel, subfamily K, member 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614720: 98297 - 143099 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509498.2 XP_003509546.1 2 - 3 639 91

View Full Product Details