Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd33
Entrez Gene ID
100765182
Gene Name
ankyrin repeat domain 33
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615454: 192707 - 208654 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512414.1 XP_003512462.1 3 - 4 435 92

View Full Product Details