Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Amigo2
Entrez Gene ID
100761994
Gene Name
adhesion molecule with Ig-like domain 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616287: 32727 - 34286 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514012.1 XP_003514060.1 1 - 1 118 85

View Full Product Details