Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fcgr2b
Entrez Gene ID
103161893
Gene Name
Fc fragment of IgG, low affinity IIb, receptor (CD32)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614301: 488567 - 523306 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647415.1 XP_007645605.1 3 - 4 231 67

View Full Product Details