Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rhbdl1
Entrez Gene ID
100771192
Gene Name
rhomboid, veinlet-like 1 (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613852: 456913 - 461074 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642212.1 XP_007640402.1 5 - 6 1121 81

View Full Product Details