Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Odf1
Entrez Gene ID
100767282
Gene Name
outer dense fiber of sperm tails 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614053: 1275637 - 1281924 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504326.2 XP_003504374.1 2 - 3 218 103

View Full Product Details