Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd37
Entrez Gene ID
100771039
Gene Name
ankyrin repeat domain 37
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614773: 27988 - 30802 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509822.2 XP_003509870.2 2 - 3 287 71

View Full Product Details