Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc30a1
Entrez Gene ID
100751596
Gene Name
solute carrier family 30 (zinc transporter), member 1
Gene Aliases
I79_018799
Chromosome Location
Chr.Un|NW_003614893: 322541 - 397237 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510495.2 XP_003510543.1 2 - 3 281 86

View Full Product Details