Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gpx8
Entrez Gene ID
100775000
Gene Name
glutathione peroxidase 8 (putative)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615347: 241804 - 245636 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512078.2 XP_003512126.1 1 - 2 452 68

View Full Product Details