Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Taf10
Entrez Gene ID
100760931
Gene Name
TAF10 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 30kDa
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615626: 91121 - 91988 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512863.2 XP_003512911.2 2 - 3 240 59
GenBank mRNA JI893058.1 - 2 - 3 240 59
JP039980.1 - 2 - 3 240 59

View Full Product Details