Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc8c
Entrez Gene ID
100765936
Gene Name
leucine rich repeat containing 8 family, member C
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614236: 129247 - 209126 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007646908.1 XP_007645098.1 1 - 2 49 76

View Full Product Details