Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vstm5
Entrez Gene ID
100752528
Gene Name
V-set and transmembrane domain containing 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614246: 805453 - 821318 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506487.2 XP_003506535.1 1 - 2 645 67

View Full Product Details