Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asb18
Entrez Gene ID
100768096
Gene Name
ankyrin repeat and SOCS box containing 18
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615970: 69202 - 131117 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513499.1 XP_003513547.1 5 - 6 1216 61

View Full Product Details