Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Exosc3
Entrez Gene ID
100753640
Gene Name
exosome component 3
Gene Aliases
I79_011915
Chromosome Location
Chr.Un|NW_003614087: 1137938 - 1142428 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645302.1 XP_007643492.1 1 - 2 59 70

View Full Product Details