Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gal3st2
Entrez Gene ID
100755837
Gene Name
galactose-3-O-sulfotransferase 2
Gene Aliases
I79_005326
Chromosome Location
Chr.Un|NW_003613726: 1407165 - 1416483 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499285.1 XP_003499333.1 2 - 3 121 72

View Full Product Details